Merck
Systemic availability and pharmacokinetics of thymol in humans.
Journal of clinical pharmacology (2002-07-03)
Claudia Kohlert, Gernot Schindler, Reinhard W März, Gudrun Abel, Benno Brinkhaus, Hartmut Derendorf, Eva-Ulrike Gräfe, Markus Veit
ABSTRACT
Essential oil compounds such as found in thyme extract are established for the therapy of chronic and acute bronchitis. Various pharmacodynamic activities for thyme extract and the essential thyme oil, respectively, have been demonstrated in vitro, but availability of these compounds in the respective target organs has not been proven. Thus, investigation of absorption, distribution, metabolism, and excretion are necessary to provide the link between in vitro effects and in vivo studies. To determine the systemic availability and the pharmacokinetics of thymol after oral application to humans, a clinical trial was carried out in 12 healthy volunteers. Each subject received a single dose of a Bronchipret TP tablet, which is equivalent to 1.08 mg thymol. No thymol could be detected in plasma or urine. However, the metabolites thymol sulfate and thymol glucuronide were found in urine and identified by LC-MS/MS. Plasma and urine samples were analyzed after enzymatic hydrolysis of the metabolites by headspace solid-phase microextraction prior to GC analysis and flame ionization detection. Thymol sulfate, but not thymol glucuronide, was detectable in plasma. Peak plasma concentrations were 93.1+/-24.5 ng ml(-1) and were reached after 2.0+/-0.8 hours. The mean terminal elimination half-life was 10.2 hours. Thymol sulfate was detectable up to 41 hours after administration. Urinary excretion could be followed over 24 hours. The amount of both thymol sulfate and glucuronide excreted in 24-hour urine was 16.2%+/-4.5% of the dose.
Sigma-Aldrich
Thymol, FCC, FG
Pricing and availability is not currently available.
Thymol, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Thymol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Sigma-Aldrich
Thymol, meets analytical specification of Ph. Eur., BP, NF, 99-101%
Pricing and availability is not currently available.
Sigma-Aldrich
Thymol, ≥98.5%
Pricing and availability is not currently available.
Supelco
Thymol, analytical standard
Pricing and availability is not currently available.