Merck
  • Extracellular matrix components are altered in the hippocampus, cortex, and cerebrospinal fluid of patients with mesial temporal lobe epilepsy.
Extracellular matrix components are altered in the hippocampus, cortex, and cerebrospinal fluid of patients with mesial temporal lobe epilepsy.
Epilepsia (2002-07-18)
S R Perosa, M A Porcionatto, A Cukiert, J R M Martins, D Amado, H B Nader, E A Cavalheiro, J P Leite, M G Naffah-Mazzacoratti
ABSTRACT
This work studied the profile of glycosaminoglycans (GAGs) in the hippocampus, cortex, and cerebrospinal fluid of patients with temporal lobe epilepsy (TLE). The GAGs were analyzed by agarose gel electrophoresis, enzymatic degradation, and enzyme-linked immunosorbent assay (ELISA). The hippocampus of TLE patients showed increased levels of chondroitin sulfate and hyaluronic acid against normal levels of these GAGs in the neocortex and cerebrospinal fluid (CSF). These results suggest that these matrix components could be involved in the pathophysiology of TLE.
Product Description
SAFC
L-Aspartic acid
SAFC
L-Tyrosine
SAFC
L-Threonine
SAFC
L-Valine
Sigma-Aldrich
L-Tyrosine, FG
Sigma-Aldrich
L-Glutamine solution, 200 mM, Hybri-Max, sterile-filtered, suitable for hybridoma
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Leucine, 99%, FG
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Leucine, BioUltra, ≥99.5% (NT)
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Leucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Alanine, BioUltra, ≥99.5% (NT)
Supelco
L-Alanine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, ≥98% (TLC)
Sigma-Aldrich
L-Alanine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material