Merck
  • Alteration of 8-hydroxyguanosine concentrations in the cerebrospinal fluid and serum from patients with Parkinson's disease.
Alteration of 8-hydroxyguanosine concentrations in the cerebrospinal fluid and serum from patients with Parkinson's disease.
Neuroscience letters (2002-12-25)
Takashi Abe, Chiaki Isobe, Takahiko Murata, Chigumi Sato, Hideo Tohgi
ABSTRACT
In order to investigate the possible role of oxidative RNA damage in the pathogenesis of Parkinson's disease (PD), the concentrations of the oxidative stress marker 8-hydroxyguanosine (8-OHG) were measured in the cerebrospinal fluid (CSF) and the serum of patients with PD and control subjects. The concentration of 8-OHG in CSF in PD patients was approximately three-fold that in controls (P < 0.001). The concentration of 8-OHG in CSF decreased significantly with the duration of disease (r(s) = -0.46, P < 0.05). However, the concentration of 8-OHG in serum was not significantly altered in PD patients compared to that in controls. In addition, the concentration of 8-OHG in CSF showed no correlation with that in serum in both the controls and PD patients suggesting that the 8-OHG concentrations in the CSF do not reflect those in serum and may be probably reflect those in brain tissue. These in vivo findings suggest a possible role of 8-OHG and increased oxidative RNA damage in the early stage of the development of PD.
Sigma-Aldrich
Manganese, powder, ≥99.9% trace metals basis
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Molybdenum, nanopowder, <100 nm particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 2.0 mm, 99.7% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Nickel, nanopowder, <100 nm avg. part. size, ≥99% trace metals basis
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Tin, ≥99%, powder
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)