Merck
  • Certification of fat-soluble vitamins, carotenoids, and cholesterol in human serum: Standard Reference Material 968b.
Certification of fat-soluble vitamins, carotenoids, and cholesterol in human serum: Standard Reference Material 968b.
Analytical and bioanalytical chemistry (1996-08-01)
J B Thomas, M C Kline, S B Schiller, P M Ellerbe, L T Sniegoski, D L Duewer, K E Sharpless
ABSTRACT
In Standard Reference Material 968b, fat-soluble vitamins and cholesterol in human serum, certified values are provided for cholesterol, retinol, retinyl palmitate, alpha-tocopherol, trans-beta-carotene, total beta-carotene ( trans plus cis isomers), total alpha-carotene, and lutein. Non-certified values are also reported for gamma-tocopherol (includes beta-tocopherol), delta-tocopherol, zeaxanthin, beta-cryptoxanthin, trans-lycopene, trans-lycopene, trans-alpha-carotene, total lycopene, 9- cis-betacarotene, 13- plus 15- cis-beta-carotene, and 15- cis-beta-carotene. Both certified and non-certified values are based on the agreement among results from three different liquid chromatographic analytical procedures developed at NIST and from an interlaboratory comparison exercise among institutions that participate in a NIST-managed Micronutrients Measurement Quality Assurance Program. Cholesterol is certified in this material using the NIST isotope dilution/mass spectrometric definitive method.
Product Description
Supelco
Retinol palmitate, analytical standard
Sigma-Aldrich
α-Carotene, ≥95.0% (HPLC)
Supelco
Retinyl Palmitate (Vitamin A Palmitate), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinyl palmitate, potency: ≥1,700,000 USP units per g
Sigma-Aldrich
(±)-α-Carotene, ≥95.0% (HPLC)
Sigma-Aldrich
Retinyl palmitate, Type IV, ~1,800,000 USP units/g, oil