Merck
  • Neopterin concentrations in pediatric human immunodeficiency virus infection as predictor of disease activity.
Neopterin concentrations in pediatric human immunodeficiency virus infection as predictor of disease activity.
The Pediatric infectious disease journal (1992-04-01)
M Ellaurie, T Calvelli, A Rubinstein
ABSTRACT
Neopterin concentrations in 50 children with human immunodeficiency virus infection were correlated with disease course. The neopterin concentrations ranged from 4 to 70 nM with a mean of 34.7 +/- 25.1 (SD) nM compared with a mean of 6.1 +/- 1.6 nM in the human immunodeficiency virus-negative control group. Elevated neopterin concentrations above the upper range of the control group were detected as early as 5 months of age. Nineteen of 20 patients (95%) with neopterin concentrations above 20 nM either died or have severe clinical disease. Increasing neopterin concentrations were also associated with poor prognosis even though the first value was below 20 nM. Conversely neopterin concentrations less than 20 nM or declining concentrations were associated with a stable clinical course, except in terminal illness. With stepwise logistic regression analysis neopterin concentrations were predictive for disease progression. This predictive value was further improved by the addition of CD4 cell counts.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cysteine, 97%
SAFC
L-Cysteine
Supelco
Butyric acid, analytical standard
Sigma-Aldrich
Butyric acid, ≥99%, FG
Sigma-Aldrich
Butyric acid, ≥99%
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Butyric acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
2,3-Pyridinedicarboxylic acid, 99%
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cysteine, Wacker Chemie AG, ≥98.0%