Merck
  • Synthesis of a biotin derivative of iberiotoxin: binding interactions with streptavidin and the BK Ca2+-activated K+ channel expressed in a human cell line.
Synthesis of a biotin derivative of iberiotoxin: binding interactions with streptavidin and the BK Ca2+-activated K+ channel expressed in a human cell line.
Bioconjugate chemistry (2006-05-18)
Jon-Paul Bingham, Shumin Bian, Zhi-Yong Tan, Zoltan Takacs, Edward Moczydlowski
ABSTRACT
Iberiotoxin (IbTx) is a scorpion venom peptide that inhibits BK Ca2+-activated K+ channels with high affinity and specificity. Automated solid-phase synthesis was used to prepare a biotin-labeled derivative (IbTx-LC-biotin) of IbTx by substitution of Asp19 of the native 37-residue peptide with N--(D-biotin-6-amidocaproate)-L-lysine. Both IbTx-LC-biotin and its complex with streptavidin (StrAv) block single BK channels from rat skeletal muscle with nanomolar affinity, indicating that the biotin-labeled residue, either alone or in complex with StrAv, does not obstruct the toxin binding interaction with the BK channel. IbTx-LC-biotin exhibits high affinity (KD = 26 nM) and a slow dissociation rate (koff = 5.4 x 10(-4) s(-1)) in a macroscopic blocking assay of whole-cell current of the cloned human BK channel. Titration of IbTx-LC-biotin with StrAv monitored by high performance size exclusion chromatography is consistent with a stoichiometry of two binding sites for IbTx-LC-biotin per StrAv tetramer, indicating that steric interference hinders simultaneous binding of two toxin molecules on each of the two biotin-binding faces of StrAv. In combination with fluorescent conjugates of StrAv or anti-biotin antibody, IbTx-LC-biotin was used to image the surface distribution of BK channels on a transfected cell line. Fluorescence microscopy revealed a patch-like surface distribution of BK channel protein. The results support the feasibility of using IbTx-LC-biotin and similar biotin-tagged K+ channel toxins for diverse applications in cellular neurobiology. .
Product Description
Biotin, European Pharmacopoeia (EP) Reference Standard
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, meets USP testing specifications
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material