Merck
  • Effect of estradiol versus estradiol and testosterone on brain-activation patterns in postmenopausal women.
Effect of estradiol versus estradiol and testosterone on brain-activation patterns in postmenopausal women.
Menopause (New York, N.Y.) (2006-06-01)
Johanna S Archer, Tracy E Love-Geffen, Kathryn L Herbst-Damm, David A Swinney, Jeffrey R Chang
ABSTRACT
To determine the effect of estradiol and testosterone on brain-activation patterns in surgically postmenopausal women viewing erotic video clips using functional magnetic resonance imaging. Six women, who had undergone a bilateral oophorectomy and hysterectomy for benign disease, viewed erotic and neutral videos during functional magnetic resonance imaging while not on hormone therapy, while on estradiol therapy, and while on estradiol and testosterone therapy. Five similarly aged premenopausal women viewed the same videos. Areas of brain activation between the functional magnetic resonance imaging scans of both groups of women were compared with determine whether agonadal serum levels of sex hormones and administration of estradiol and testosterone impacted brain patterns of sexual arousal. When compared with premenopausal women, untreated postmenopausal women had significantly decreased areas of brain activation during both erotic and neutral stimulations. Administration of estradiol increased global brain-activation patterns during both visual stimulations, with erotic video viewing causing a limited increase in limbic system activation. Combined estradiol and testosterone therapy was associated with a greater activation of the central nervous system, with more limbic system activated during the erotic video. Brain-activation patterns of the postmenopausal women were similar to the premenopausal group only during the estradiol and testosterone treatment phase. Agonadal serum hormone levels result in globally decreased brain-activation patterns in postmenopausal women while viewing neutral and erotic videos. Administration of both estradiol and testosterone increase global brain activation, and both sex steroids are independently associated with enhanced limbic system response during erotic visual stimulation.
Sigma-Aldrich
β-Estradiol, BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Testosterone, purum, ≥99.0% (HPLC)
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture
Supelco
Testosterone, VETRANAL®, analytical standard
Testosterone, European Pharmacopoeia (EP) Reference Standard
Supelco
Testosterone solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
β-Estradiol, analytical standard
Supelco
17β-Estradiol solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®