Merck
  • Relationship between omega-3 fatty acids and plasma neuroactive steroids in alcoholism, depression and controls.
Relationship between omega-3 fatty acids and plasma neuroactive steroids in alcoholism, depression and controls.
Prostaglandins, leukotrienes, and essential fatty acids (2006-09-09)
L R G Nieminen, K K Makino, N Mehta, M Virkkunen, H Y Kim, J R Hibbeln
ABSTRACT
Deficiency in the long-chain omega-3 fatty acid, docosahexaenoic acid (DHA) has been associated with increased corticotropin releasing hormone and may contribute to hypothalamic pituitary axis (HPA) hyperactivity. Elevated levels of the neuroactive steroids, allopregnanolone (3alpha,5alpha-THP) and 3alpha,5alpha-tetrahydrodeoxycorticosterone (THDOC) appear to counter-regulate HPA hyperactivity. Plasma essential fatty acids and neurosteroids were assessed among 18 male healthy controls and among 34 male psychiatric patients with DSM-III alcoholism, depression, or both. Among all subjects, lower plasma DHA was correlated with higher plasma THDOC (r = -0.3, P < 0.05) and dihydroprogesterone (DHP) (r = -0.52, P < 0.05). Among psychiatric patients lower DHA was correlated with higher DHP (r = -0.60, P < 0.01), and among healthy controls lower plasma DHA was correlated with higher THDOC (r = -0.83, P < 0.01) and higher isopregnanolone (3beta,5alpha-THP) (r = -0.55, P < 0.05). In this pilot observational study, lower long-chain omega-3 essential fatty acid status was associated with higher neuroactive steroid concentrations, possibly indicating increased feedback inhibition of the HPA axis.
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Nicotinic acid, meets USP testing specifications
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Nicotinic acid, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinic acid, ≥98%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Homovanillic acid, Fluorimetric reagent
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Acetone, analytical standard
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Methanol, analytical standard
Supelco
Nicotinic acid, analytical standard
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
5α-Dihydrotestosterone (DHT) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®