Merck
The human cerebrospinal fluid metabolome.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-05-27)
David S Wishart, Michael J Lewis, Joshua A Morrissey, Mitchel D Flegel, Kevin Jeroncic, Yeping Xiong, Dean Cheng, Roman Eisner, Bijaya Gautam, Dan Tzur, Summit Sawhney, Fiona Bamforth, Russ Greiner, Liang Li
ABSTRACT
With continuing improvements in analytical technology and an increased interest in comprehensive metabolic profiling of biofluids and tissues, there is a growing need to develop comprehensive reference resources for certain clinically important biofluids, such as blood, urine and cerebrospinal fluid (CSF). As part of our effort to systematically characterize the human metabolome we have chosen to characterize CSF as the first biofluid to be intensively scrutinized. In doing so, we combined comprehensive NMR, gas chromatography-mass spectrometry (GC-MS) and liquid chromatography (LC) Fourier transform-mass spectrometry (FTMS) methods with computer-aided literature mining to identify and quantify essentially all of the metabolites that can be commonly detected (with today's technology) in the human CSF metabolome. Tables containing the compounds, concentrations, spectra, protocols and links to disease associations that we have found for the human CSF metabolome are freely available at http://www.csfmetabolome.ca.
Product Description
Supelco
L-Alanine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
SAFC
L-Glutamine Solution 200 mM, 29.23 mg/mL in saline, solution, suitable for cell culture
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Mannitol, European Pharmacopoeia (EP) Reference Standard
Alanine, European Pharmacopoeia (EP) Reference Standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Leucine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethyl sulfone, qNMR standard, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Alanine, ≥99%
Sigma-Aldrich
Dimethylamine, anhydrous, ≥99%
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Valine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
myo-Inositol, ≥99% (GC), BioReagent
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Supelco
Methanol, analytical standard
Millipore
D-(+)-Mannose, suitable for microbiology, ≥99%
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)
SAFC
L-Aspartic acid
Supelco
Acetone, analytical standard
Millipore
D-(+)-Mannose, ≥99.0% (sum of enantiomers, HPLC), suitable for microbiology