- Tin and the thymus gland: a review.
ABSTRACT
Experimental studies over the last decade have suggested an association between thymus immune and homeostatic function and exogenous tin. It has been hypothesized that the thymus gland synthesizes and secretes one or more tin bearing factors that enhance immune defenses against malignancy and retard the gradual loss of immune capacity with senescence. This review conciliates data from several divergent areas of research in order to explore the rationale for the above concepts.
Product Description
Tin, foil, light tested, 50x50mm, thickness 0.008mm, 99.75%
Tin, foil, not light tested, 150x150mm, thickness 0.006mm, 97.4%
Tin, foil, not light tested, 150x150mm, thickness 0.009mm, 97.4%
Tin, foil, not light tested, 150x150mm, thickness 0.007mm, 97.4%
Tin, wire reel, 5m, diameter 0.25mm, as drawn, 99.99+%
Tin, foil, not light tested, 25x25mm, thickness 0.007mm, 99.75%
Tin, foil, not light tested, 25x25mm, thickness 0.006mm, 99.75%
Tin, foil, 10m coil, thickness 0.025mm, as rolled, 98.8%
Tin, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 10mm disks, thickness 0.006mm, 97.4%
Tin, foil, 0.2m coil, thickness 0.06mm, coil width 73mm, as rolled, 99.99+%
Tin, foil, 50x50mm, thickness 0.1mm, as rolled, 98.8%
Tin, foil, 10m coil, thickness 0.007mm, 97.4%
Tin, foil, 0.5m coil, thickness 0.0125mm, 97.4%
Tin, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%
Tin, foil, 10m coil, thickness 0.084mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 100x100mm, thickness 0.65mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 2.0mm, as rolled, 99.99+%
Tin, foil, 300x300mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 300x300mm, thickness 0.5mm, as rolled, 98.8%
Tin, foil, 0.5m coil, thickness 0.01mm, 97.4%
Tin, foil, 0.5m coil, thickness 0.125mm, as rolled, 98.8%