Merck
  • Enterobacter asburiae sp. nov., a new species found in clinical specimens, and reassignment of Erwinia dissolvens and Erwinia nimipressuralis to the genus Enterobacter as Enterobacter dissolvens comb. nov. and Enterobacter nimipressuralis comb. nov.
Enterobacter asburiae sp. nov., a new species found in clinical specimens, and reassignment of Erwinia dissolvens and Erwinia nimipressuralis to the genus Enterobacter as Enterobacter dissolvens comb. nov. and Enterobacter nimipressuralis comb. nov.
Journal of clinical microbiology (1986-06-01)
D J Brenner, A C McWhorter, A Kai, A G Steigerwalt, J J Farmer
ABSTRACT
Enterobacter asburiae sp. nov. is a new species that was formerly referred to as Enteric Group 17 and that consists of 71 strains, 70 of which were isolated from humans. Enterobacter asburiae sp. nov. strains gave positive reactions in tests for methyl red, citrate utilization (Simmons and Christensen's), urea hydrolysis, L-ornithine decarboxylase, growth in KCN, acid and gas production from D-glucose, and acid production from L-arabinose, cellobiose, glycerol (negative in 1 to 2 days, positive in 3 to 7 days), lactose, D-mannitol, alpha-methyl-D-glucoside, salicin, D-sorbitol, sucrose, trehalose, and D-xylose. They gave negative reactions in the Voges-Proskauer test and in tests for indole, H2S production, phenylalanine, L-lysine decarboxylase, motility, gelatin, utilization of malonate, lipase, DNase, tyrosine clearing, acid production from adonitol, D-arabitol, dulcitol, erythritol, i(myo)-inositol, melibiose, and L-rhamnose. They gave variable reactions in tests for L-arginine dihydrolase (25% positive after 2 days) and acid production from raffinose (69% positive after 2 days). Thirty-four Enterobacter asburiae sp. nov. strains were tested for DNA relatedness by the hydroxyapatite method with 32PO4-labeled DNA from the designated type strain (1497-78, ATCC 35953). The strains were 69 to 100% related in 60 degrees C reactions and 63 to 100% related in 75 degrees C reactions. Divergence within related sequences was 0 to 2.5%. Relatedness of Enterobacter asburiae sp. nov. to 84 strains of members of the Enterobacteriaceae was 5 to 63%, with closest relatedness to strains of Enterobacter cloacae, Erwinia dissolvens, Enterobacter taylorae, Enterobacter agglomerans, Erwinia nimipressuralis, and Enterobacter gergoviae. All strains tested were susceptible to gentamicin and sulfdiazine, and most were susceptible to chloramphenicol, colistin, kanamycin, nalidixic acid, carbenicillin and streptomycin. All strains were resistant to ampicillan, cephalothin, and penicillin, and most were resistant or moderately resistant to tetracycline. Enterobacter asburiae sp. nov strains were isolated from a variety of human sources, most prevalent of which were urine (16 strains), respiratory sources (15 strains), stools (12 strains), wounds (11 strains), and blood (7 strains). The clinical significance of Enterobacter aburiae is not known. As a result of this and previous studies, proposals are made to transfer Erwinia dissolvens and Erwinia nimipressuralis to the genus Enterobacter as Enterobacter dissolvens comb. nov. and Enterobacter nimipressuralis comb. nov., respectively.
Sigma-Aldrich
Adonitol, BioXtra, ≥99.0% (HPLC)
Sigma-Aldrich
Adonitol, ≥99%