Merck
Stereospecificity of the oxidation of ethanol by catalase.
The Journal of biological chemistry (1974-05-25)
R J Corrall, H M Rodman, J Margolis, B R Landau
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa