- Bile salt hydrolase activity in probiotics.
Bile salt hydrolase activity in probiotics.
Applied and environmental microbiology (2006-03-07)
Máire Begley, Colin Hill, Cormac G M Gahan
PMID16517616
Sigma-Aldrich
Choloylglycine Hydrolase from Clostridium perfringens (C. welchii), lyophilized powder, ≥100 units/mg protein