Merck
  • Nucleosidediphosphatase activity in the Golgi apparatus and its usefulness for cytological studies.
Nucleosidediphosphatase activity in the Golgi apparatus and its usefulness for cytological studies.
Proceedings of the National Academy of Sciences of the United States of America (1961-06-15)
A B NOVIKOFF, S GOLDFISCHER
Sigma-Aldrich
Thiamine pyrophosphate, ≥95%