Merck
  • [Ginsenoside Rg3 induces apoptosis of human lung squamous cell carcinoma SK-MES-1 cell line].
[Ginsenoside Rg3 induces apoptosis of human lung squamous cell carcinoma SK-MES-1 cell line].
Nan fang yi ke da xue xue bao = Journal of Southern Medical University (2009-09-26)
Xin Wang, Yu-ling Zheng, Ke Li, Na Lin, Qing-xia Fan
ABSTRACT
To investigate the effect of ginsenoside Rg3 on the apoptosis and survivin expression in human lung squamous cell carcinoma cell line SK-MES-1. SK-MES-1 cells were divided into Rg3 treatment group, blank control group and positive control (arsenic trioxide) group. The apoptotic rate of the cells in each group was determined using flow cytometry, and the expression of survivin protein and mRNA was detected by immunocytochemistry and RT-PCR, respectively. A 48-h treatment with Ginsenoside Rg3 induced increased apoptotic rate of SK-MES-1 cells in a dose-dependent manner. Ginsenoside Rg3 significantly downregulated the expressions of survivin protein and mRNA as compared with the expression levels in the blank control group (P<0.05). Ginsenoside Rg3 can induce the apoptosis of SK-MES-1 cells, the mechanism of which may involve inhibited survivin expression.
Product Description
SK MES 1, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.