Merck
  • ISOLATION AND PROPERTIES OF AN EXOCELLULAR NUCLEASE OF SERRATIA MARCESCENS.
ISOLATION AND PROPERTIES OF AN EXOCELLULAR NUCLEASE OF SERRATIA MARCESCENS.
Journal of bacteriology (1963-02-01)
G N Eaves, C D Jeffries
ABSTRACT
Eaves, George N. (Wayne State University College of Medicine, Detroit, Mich.) and Charles D. Jeffries. Isolation and properties of an exocellular nuclease of Serratia marcescens. J. Bacteriol. 85:273-278. 1963.-The exocellular nuclease of Serratia marcescens, isolated by anion-exchange chromatography on diethylaminoethyl-Sephadex, depolymerized deoxyribonucleic acid, ribonucleic acid, and the polynucleotide which is refractory to pancreatic ribonuclease activity. The enzyme was tentatively classified as a nonspecific phosphodiesterase. Magnesium was essential for activity, which was optimal at pH 8.8. The purified enzyme was completely inactivated by heating at 50 C for 15 min.
Sigma-Aldrich
Benzonase® Nuclease, ultrapure, ≥250 units/μL, ≥99% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution, ultrapure grade
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution