Merck
  • [Application of solid-phase microextraction technique to the detection of amphetamines in urine by GC].
[Application of solid-phase microextraction technique to the detection of amphetamines in urine by GC].
Fa yi xue za zhi (2003-01-23)
W Liu, M Shen
ABSTRACT
A simple and rapid detection of nine amphetamines co-existing in urine was described. In the test, the method of solid-phase micro-extraction (SPME) by GC technique was used. Urine (1.0 ml), NaCl (0.3 g) and 4-phenylbutylamine (internal standard) were added into a vial (1.5 ml), then the sample was adjusted to pH 12 with 10% NaOH and sealed with a teflon-coated septum. After immersion of the SPME fiber (100 PDME) in the sample for 15 min, the SPME needle was inserted into the injection port of the GC and extruded for 3 min. The result showed that each peak from nine amphetamines compounds and internal standard was clearly separated. The calibration curves were linear from 0.2 to 15 micrograms/ml for most of five amphetamines with r between 0.9928-0.9995. The CV were less 10%. It is concluded that the method is simple, quick, accurate and useful for the practical detection of urine concentration of amphetamines.
Product Description
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1, plain gray hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub