Merck
Tantalum foil cuffs in peripheral nerve surgery.
Surgery (1948-03-01)
E E CLIFFTON
Product Description
Tantalum, foil, light tested, 25x25mm, thickness 0.005mm, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.0025mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.009mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.004mm, 99.9%
Tantalum, foil, 10mm disks, thickness 0.034mm, as rolled, 99.9%
Tantalum, foil, 0.2m coil, thickness 0.0125mm, 99.9%
Tantalum, foil, 10m coil, thickness 0.04mm, coil width 1mm, annealed, 99.9%
Tantalum, foil, 10mm disks, thickness 0.006mm, 99.9%
Tantalum, foil, 10m coil, thickness 0.04mm, coil width 0.7mm, annealed, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.005mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.02mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.0075mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.005mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.0025mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.0075mm, 99.9%
Tantalum, foil, light tested, 150x300mm, thickness 0.05mm, annealed, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.009mm, 99.9%
Tantalum, foil, not light tested, 50x50mm, thickness 0.009mm, 99.9%
Tantalum, foil, light tested, 150x150mm, thickness 0.05mm, annealed, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.002mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.9%
Tantalum, foil, not light tested, 100x100mm, thickness 0.004mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.015mm, 99.9%
Tantalum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Tantalum, foil, light tested, 50x50mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, light tested, 100x100mm, thickness 0.075mm, annealed, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.03mm, as rolled, 99.9%
Tantalum, foil, 8mm disks, thickness 0.125mm, annealed, 99.9%
Tantalum, foil, 6mm disks, thickness 0.15mm, annealed, 99.9%
Tantalum, foil, light tested, 25x25mm, thickness 0.0075mm, 99.9%