- Mercury contamination removed with tin foil.
Mercury contamination removed with tin foil.
Operative dentistry (1984-01-01)
R Grajower, J Mann, I Lewinstein
PMID6591141
Product Description
Tin, foil, not light tested, 50x50mm, thickness 0.004mm, 99.75%
Tin, foil, light tested, 50x50mm, thickness 0.025mm, as rolled, 99.99+%
Tin, foil, light tested, 50x50mm, thickness 0.006mm, 97.4%
Tin, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99.99+%
Tin, foil, 150x150mm, thickness 0.25mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 98.8%
Tin, foil, 150x150mm, thickness 0.5mm, as rolled, 98.8%
Tin, foil, 25x25mm, thickness 0.15mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Tin, foil, 150x150mm, thickness 0.125mm, as rolled, 99.99+%
Tin, foil, 0.5m coil, thickness 1.0mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Tin, foil, 100x100mm, thickness 0.20mm, as rolled, 98.8%
Tin, foil, 10m coil, thickness 0.006mm, 97.4%
Tin, foil, 100x100mm, thickness 0.25mm, as rolled, 98.8%
Tin, foil, 10m coil, thickness 0.096mm, coil width 41mm, as rolled, 99.95%
Tin, foil, 100x100mm, thickness 2.0mm, as rolled, 98.8%
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 98.8%
Tin, foil, 25x25mm, thickness 0.25mm, as rolled, 98.8%
Tin, foil, 150x150mm, thickness 2.0mm, as rolled, 98.8%
Tin, foil, 25x25mm, thickness 1.0mm, as rolled, 98.8%
Tin, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Tin, foil, 300x300mm, thickness 0.1mm, as rolled, 98.8%
Tin, foil, 100x100mm, thickness 0.125mm, as rolled, 98.8%
Tin, foil, 50x50mm, thickness 0.15mm, as rolled, 99.99+%
Tin, foil, 25x25mm, thickness 0.5mm, as rolled, 98.8%
Tin, foil, 0.5m coil, thickness 0.025mm, as rolled, 98.8%
Tin, foil, 50x50mm, thickness 1.0mm, as rolled, 98.8%
Tin, foil, 10mm disks, thickness 0.004mm, 99.75%
Tin, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%