Merck
  • Simulation of the effect of maize porridge fortified with grain amaranth or micronutrient powder containing NaFeEDTA on iron intake and status in Kenyan children.
Simulation of the effect of maize porridge fortified with grain amaranth or micronutrient powder containing NaFeEDTA on iron intake and status in Kenyan children.
Public health nutrition (2012-12-12)
Catherine W Macharia-Mutie, Agnes M Omusundi, John M Mwai, Alice M Mwangi, Inge D Brouwer
ABSTRACT
Simulating the probable impact of grain amaranth and highly absorbable, low-Fe micronutrient powder (MNP) on Fe status in a potential target population is an essential step in choosing and developing an appropriate actual intervention. We simulated the potential effect of fortifying maize porridge with grain amaranth or MNP on the prevalence of inadequate Fe intake and Fe deficiency using data from two cross-sectional surveys. In the first survey (2008), dietary intake data were collected by two 24 h recalls (n 197). Biochemical data (n 70) were collected in the second survey (2010). A simulation with daily consumption for 80 d of non-fortified maize porridge (60 g of maize flour), amaranth-enriched porridge (80 g of grain amaranth–maize flour, 70:30 ratio) or maize porridge fortified with MNP (2.5mg Fe as NaFeEDTA) was done. Mwingi District, Kenya. Pre-school children aged 12–23 months. Prevalence of anaemia, Fe deficiency and Fe-deficiency anaemia was 49 %, 46% and 24 %, respectively. Consumption of non-fortified, amaranth-enriched and MNP-fortified maize porridge was estimated to provide a median daily Fe intake of 8.6 mg, 17.5mg and 11.1 mg, respectively. The prevalence of inadequate Fe intake was reduced to 35% in the amaranth-enriched porridge group and 45% in the MNP-fortified porridge group, while ferritin concentration was increased in both (by 1.82 (95% CI 1.42, 2.34) mg/l and 1.80 (95% CI 1.40, 2.31) μg/l, respectively; P,0.005) compared with the non-fortified maize porridge group, resulting in a decreased prevalence of Fe deficiency (27 %) in the two fortification groups. Addition of grain amaranth or low-Fe MNP to maize-based porridge has potential to improve Fe intake and status in pre-school children.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid ferric sodium salt, suitable for plant cell culture, BioReagent, crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid iron(III) sodium salt, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected