Merck
Evaluation of cutaneous palpebral anthrax.
Cutaneous and ocular toxicology (2013-04-10)
Recep Tekin, Seyhmus Ari, Tuba Dal, Safak Kaya, Mehmet Zeki Kortak, Birgül Dursun, Saim Dayan
ABSTRACT
Anthrax is a rare disease caused by Bacillus anthracis. Antrax is zoonotic disease and is often encountered in persons engaged in animal husbandry. Cutaneous anthrax is approximately 95% of anthrax in humans. Palbebral involvement is rare. In this study, we aimed to evaluate the clinical presentation, diagnosis and treatment of cases with cutaneous palpebral anthrax. In this study, the patients diagnosed of cutaneous palpebral anthrax between January 2000 and December 2012, were investigated and evaluated, retrospectively. Cutaneous palpebral anthrax was diagnosed by the presence of typical anthrax lesion and/or observation of gram-positive encapsulated bacilli in gram prepations and/or culture positive of samples taken from lesions. In the cases who were culture-negative and without bacilli in gram-staining, the diagnosis was based on the presence of characteristic clinical presentation with a history of severe scarring formation, swelling, black eschar and positive response to the treatment. A total of 21 patients with cutaneous palpebral anthrax admitted to the two hospitals between January 2000 and December 2012. Eight patients were male (38.1%) and 13 patients were female (61.9%), and the mean age was 31 ± 21.2 (range 1-82 years). The most common symptoms on admission to the hospital were swelling and redness on the skin. Periorbital lesions were in the right eye in 14 cases and the most common eyelid involvement was seen in upper eyelid with 15 cases. The diagnosis was based on isolation of bacteria in five (23.8%) cases, detection of gram-positive bacilli in direct examination of characteristic lesion material in six (28.5%) cases. Ten (47.7%) cases were diagnosed by the characteristic appearance of the lesion. Malignant pustule was seen in all of our patients and seven cases (33.4%) had malignant edema. In the treatment, penicilin was used for 10 (47.7%) cases, ampicillin-sulbactam for five (23.8%) cases and, ciprofloxacin for three (14.3%) cases. Cicatricial ectropion was observed in 10 (47.7%) patients, lagophthalmos developed in four (19%) patients, and corneal scar in two (9.5%) patients. The distribution of the cases did not differ by the year but showed a density in the months from July to September (62.7%). Early diagnosis and high dose antibiotic treatment can facilitate the treatment and prevent development of eyelid complications including cicatricial ectropion, corneal scars and palpebral symphysis. Prolonged follow-up is necessary in patients who develop complications and surgical intervention.
Product Description
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ampicillin trihydrate, VETRANAL®, analytical standard
Sigma-Aldrich
Ampicillin trihydrate, 900-1050 μg/mg anhydrous basis (HPLC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Sulbactam
Sulbactam sodium, European Pharmacopoeia (EP) Reference Standard
Sulbactam, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Cefazolin sodium salt, 89.1-110.1%
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Ampicillin sodium salt
Sigma-Aldrich
Ampicillin sodium salt, BioXtra, suitable for cell culture
Sigma-Aldrich
Ampicillin sodium salt, powder or crystals, BioReagent, suitable for cell culture
Sultamicillin for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Ampicillin
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sultamicillin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard