Merck
  • GidA expression in Salmonella is modulated under certain environmental conditions.
GidA expression in Salmonella is modulated under certain environmental conditions.
Current microbiology (2013-04-13)
Jaclyn M Rehl, Daniel C Shippy, Nicholas M Eakley, Megan D Brevik, Jordan M Sand, Mark E Cook, Amin A Fadl
ABSTRACT
Glucose-inhibited division (GidA) protein is widely distributed in nature, and is highly conserved among bacteria and eukarya. In our previous study, a gidA mutant was attenuated in both in vitro and in vivo models of Salmonella infection. Furthermore, deletion of gidA resulted in a marked reduction in the expression of many virulence genes and proteins, suggesting a role for GidA in the regulation of Salmonella virulence. In this study, the effect of different environmental conditions (glucose, EDTA, and pH 5) on GidA expression in Salmonella was examined. Transcriptional analysis using real-time RT-PCR and a β-galactosidase assay, displayed no differences in gidA transcription and promoter activity in different environmental conditions. Conversely, semiquantitative Western blot analysis revealed a significant increase in the GidA expression in Salmonella when grown under different environmental conditions. Salmonella in vitro virulence assays showed an increased virulence potential in the environmental conditions correlating to the increase in GidA expression. Together, our data indicate that GidA expression is modulated under different environmental conditions which correlate to increased Salmonella in vitro virulence.
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder