Merck
  • Percentage exposure of root dentin collagen after application of two irrigation protocols with manual or rotary instrumentation and two methacrylate resin-based sealers.
Percentage exposure of root dentin collagen after application of two irrigation protocols with manual or rotary instrumentation and two methacrylate resin-based sealers.
The journal of adhesive dentistry (2013-04-18)
Santiago González-López, Ernesto Martín-Altuve, Victoria Bolaños-Carmona, Purificación Sánchez-Sánchez, Alejandro Rodríguez-Navarro
ABSTRACT
To compare the percentage of collagen exposed in dentin root thirds after two irrigation protocols with manual or rotary instrumentation using two methacrylate resin-based sealers. Forty-eight single-root human teeth were prepared with manual (n = 24) or nickeltitanium ProFile rotary (n = 24) instrumentation, using 5% NaOCl between instruments and 5 ml 17% EDTA as final irrigant or 20% citric acid + 2% chlorhexidine (CHX) between instruments and as the final irrigant. RealSeal or EndoREZ were used as filling materials. One 1-mm slice per third was abraded and stained with Masson's trichrome method. Mean exposed collagen values were obtained in four areas from each section (at 60X magnification) and a complete factorial ANOVA was used to analyze the influence of the study variables. Non-parametric Mann-Whitney's test was used to compare groups. Differences with p < 0.05 were considered significant. A significantly higher percentage of collagen was exposed in all thirds with the use of the 20% citric acid + 2% CHX protocol with rotary vs manual instrumentation, but percent collagen exposed did not differ as a function of the filling material. After the 5% NaOCl + 17% EDTA protocol, the percentage of collagen exposed did not differ between rotary and manual instrumentation but was higher with the use of RealSeal. The highest percentage exposure of collagen was with 20% citric acid + 2% CHX using rotary instrumentation, regardless of the filling material.
Product Description
Supelco
Ethylenediaminetetraacetic acid disodium salt dihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, BioUltra, 98.5-101.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Eosin Y disodium salt, ≥85 % by HPLC
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt dihydrate, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Ethylenediaminetetraacetic acid calcium disodium salt hydrate
Supelco
Citric acid, Anhydrous, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, 2%, solution
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Supelco
Citric acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland