Merck
  • Assessing bioavailability of DDT and metabolites in marine sediments using solid-phase microextraction with performance reference compounds.
Assessing bioavailability of DDT and metabolites in marine sediments using solid-phase microextraction with performance reference compounds.
Environmental toxicology and chemistry (2013-05-11)
Lian-Jun Bao, Fang Jia, J Crago, Eddy Y Zeng, D Schlenk, Jay Gan
ABSTRACT
Solid-phase microextraction (SPME) has often been used to estimate the freely dissolved concentration (Cfree ) of organic contaminants in sediments. A significant limitation in the application of SPME for Cfree measurement is the requirement for attaining equilibrium partition, which is often difficult for strongly hydrophobic compounds such as DDT. A method was developed using SPME with stable isotope-labeled analogues as performance reference compounds (PRCs) to measure Cfree of DDT and metabolites (DDTs) in marine sediments. Six (13) C-labeled or deuterated PRCs were impregnated into polydimethylsiloxane (PDMS) fiber before use. Desorption of PRCs from PDMS fibers and absorption of DDTs from sediment were isotropic in a range of sediments evaluated ex situ under well-mixed conditions. When applied to a historically contaminated marine sediment from a Superfund site, the PRC-SPME method yielded Cfree values identical to those found by using a conventional equilibrium SPME approach (Eq-SPME), whereas the time for mixing was reduced from 9 d to only 9 h. The PRC-SPME method was further evaluated against bioaccumulation of DDTs by Neanthes arenaceodentata in the contaminated sediment with or without amendment of activated carbon or sand. Strong correlations were consistently found between the derived equilibrium concentrations on the fiber and lipid-normalized tissue residues for DDTs in the worms. Results from the present study clearly demonstrated the feasibility of coupling PRCs with SPME sampling to greatly shorten sampling time, thus affording much improved flexibility in the use of SPME for bioavailability evaluation.
Product Description
Supelco
4,4′-DDT, PESTANAL®, analytical standard
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Deuterium, 99.9 atom % D
Supelco
4,4′-DDT solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Sigma-Aldrich
Deuterium, 99.8 atom % D