Merck
  • Participation of citral in the bronchodilatory effect of ginger oil and possible mechanism of action.
Participation of citral in the bronchodilatory effect of ginger oil and possible mechanism of action.
Fitoterapia (2013-05-21)
Thitiya Mangprayool, Sajeera Kupittayanant, Nuannoi Chudapongse
ABSTRACT
The extract of ginger, the rhizomes of Zingiber officinale Roscoe (Zingiberaceae), has been reported to possess anti-hyperactivity and anti-inflammation on airway. The present study described brochodilatory activity of ginger oil and identified its active compound. Ginger oil was extracted by hydro-distillation. The compositions of ginger oil were analyzed by gas chromatography and mass spectrometer. Citral, eucalyptol and camphene were found to be the major components. Ginger oil and citral, but not camphene, suppressed rat tracheal contraction induced by carbachol (CCh). Consistent with previous report, eucalyptol showed a relaxing effect on rat airway. Since the content of eucalyptol in ginger oil was relatively low, the contribution of eucalyptol to the bronchodilatory effect of ginger oil was small. To elucidate the mechanisms responsible for the myorelaxing effect, propranolol (a β-adrenergic receptor antagonist), indomethacin (a COX inhibitor) and L-NAME (a NOS inhibitor) were used to block the inhibitory effects of ginger oil and citral. It was found that propranolol, but not indomethacin and L-NAME, reversed bronchodilatory effects of both ginger oil and citral, suggesting that a possible mechanism involved β-adrenergic receptor. This study provides the pharmacological basis supporting the therapeutic potential of Z. officinale rhizomes as a bronchodilator.
Product Description
Supelco
1,8-Cineole, primary reference standard
Sigma-Aldrich
Camphene, 95%
Supelco
Camphene, analytical standard
Supelco
Propranolol hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Citral, natural, ≥96%, FCC, FG
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
Sigma-Aldrich
Citral, 95%
Sigma-Aldrich
Citral, mixture of cis and trans, ≥96%, FG
Sigma-Aldrich
Eucalyptol, 99%
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Supelco
Eucalyptol, analytical standard
Supelco
Propranolol hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Propranolol hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Cineole, European Pharmacopoeia (EP) Reference Standard
Carbachol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.