Merck
  • A highly selective and sensitive ratiometric chemodosimeter for Hg2+ ions based on an iridium(III) complex via thioacetal deprotection reaction.
A highly selective and sensitive ratiometric chemodosimeter for Hg2+ ions based on an iridium(III) complex via thioacetal deprotection reaction.
Dalton transactions (Cambridge, England : 2003) (2013-05-21)
Fengniu Lu, Masaki Yamamura, Tatsuya Nabeshima
ABSTRACT
A novel luminescent biscyclometalated iridium(III) complex [Ir(C^N)2bpy]PF6 (Ir-S, bpy = 2,2'-bipyridine) containing two 2-phenylpyridine (ppy) cyclometalating ligands (C^N) functionalized with 1,3-dithiane for the detection of Hg(2+) ions has been synthesized and characterized by spectroscopic and photophysical measurements. The luminescence of Ir-S exhibits a ratiometric response upon the addition of Hg(2+) ions. The absorption, emission, (1)H NMR and ESI mass spectral changes of Ir-S in the absence and presence of Hg(2+) ions have all demonstrated the Hg(2+)-promoted thioacetal deprotection reaction of Ir-S and the generation of a complex [Ir(pba)2bpy]PF6 (Ir-CHO, Hpba = 4-(pyridin-2-yl)benzaldehyde). DFT calculation studies suggest that the dominant participation of the 1,3-dithiane group in the HOMO of Ir-S leads to different excited states and distinct excited energies of Ir-S and Ir-CHO and consequently results in their different emission properties. The titration and competition experiments significantly reveal the highly sensitive and selective properties of Ir-S as a promising indicator for Hg(2+) ions over other metal cations.
Sigma-Aldrich
Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis
Sigma-Aldrich
Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%
Sigma-Aldrich
Iridium, powder, 99.9% trace metals basis
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Supelco
Benzaldehyde, analytical standard
Sigma-Aldrich
Benzaldehyde, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Benzaldehyde, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzaldehyde, Pharmaceutical Secondary Standard; Certified Reference Material