Merck
  • Partition, sorption and structure activity relation study of dialkoxybenzenes that modulate insect behavior.
Partition, sorption and structure activity relation study of dialkoxybenzenes that modulate insect behavior.
Chemosphere (2013-06-05)
Parisa Ebrahimi, Jacob Spooner, Noham Weinberg, Erika Plettner
ABSTRACT
Some dialkoxybenzenes are promising new insect control agents. These compounds mimic naturally occurring odorants that modulate insect behavior. Before applying these compounds, however, their persistence and biodegradability at the application site and in the environment should be understood. The fate of organic compounds in the environment is a complex phenomenon which is influenced by many processes such as sorption to soil components, sedimentation, volatilization, and uptake by plants, as well as biotic and abiotic chemical degradation. In this study, the octanol-water partition coefficient, volatility and sorption on soil components (sand, clay and organic matter) of selected dialkoxybenzenes as well as structure activity relationships with regard to partition, volatility and sorption were investigated. Additionally, calculations of partition, molar volume and molecular surface areas were done, to understand structure-activity relationships of the physical properties.
Product Description
Supelco
Benzene solution, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzene, analytical standard
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Supelco
Benzene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzene, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Benzene, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzene, suitable for HPLC, ≥99.9%