Merck
  • The preferential selection of K65R in HIV-1 subtype C is attenuated by nucleotide polymorphisms at thymidine analogue mutation sites.
The preferential selection of K65R in HIV-1 subtype C is attenuated by nucleotide polymorphisms at thymidine analogue mutation sites.
The Journal of antimicrobial chemotherapy (2013-06-12)
Cédric F Invernizzi, Dimitrios Coutsinos, Maureen Oliveira, Rita S Schildknecht, Hongtao Xu, Simani Gaseitsiwe, Daniela Moisi, Bluma G Brenner, Mark A Wainberg
ABSTRACT
We recently reported the preferential selection of the K65R resistance mutation in subtype C HIV-1 compared with subtype B and showed the underlying mechanism to be dependent on subtype C-specific silent nucleotide polymorphisms, i.e. genomic mutations that change the genotype but not the phenotype. The number of clinical reports demonstrating elevated numbers of K65R nevertheless suggests the existence of factors limiting the increased incidence of K65R mutations. Thus, we investigated the contributions of subtype C-specific silent nucleotide polymorphisms at thymidine analogue mutation (TAM) sites 70, 210 and/or 219 that might reduce the previously described preferential selection of K65R in subtype C HIV-1 associated with subtype C-specific nucleotide polymorphisms at sites 64/65. Cell culture drug selections were performed with various drugs in MT2 cells. The use of nucleoside/nucleotide reverse transcriptase inhibitors [N(t)RTIs] as single drugs or in combination confirmed the more frequent selection of K65R by multiple N(t)RTIs in a subtype B virus that contained the 64/65 nucleotide polymorphisms of subtype C than in a wild-type subtype B virus. This effect was attenuated in the presence of several silent TAM nucleotide polymorphisms, except when stavudine was employed in the selection protocol. These results further demonstrate that stavudine can preferentially select for K65R in subtype C virus and also provide a basis for understanding the importance of silent nucleotide polymorphisms in regard to altered HIV drug resistance profiles.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Thymidine, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.