Merck
  • Sodium bicarbonate use in shock and cardiac arrest: attitudes of pediatric acute care physicians.
Sodium bicarbonate use in shock and cardiac arrest: attitudes of pediatric acute care physicians.
Critical care medicine (2013-06-14)
Melissa J Parker, Christopher S Parshuram
ABSTRACT
To evaluate the preferences and self-reported practices of pediatric acute care physicians with respect to sodium bicarbonate administration to infants and children in shock or cardiac arrest. National survey study utilizing a self-administered questionnaire. Thirteen Canadian pediatric tertiary care centers. Canadian pediatric critical care physicians, pediatric emergency physicians, and trainees in these subspecialties. None. Survey items were evaluated based on Yes/No responses, frequency responses, and Likert scales. Overall response rate was 53% (151/284) with 49.0% (74/151) citing pediatric critical care as their primary practice. 82.0% of respondents (123/150) indicated they would administer sodium bicarbonate as part of ongoing resuscitation for septic shock, whereas 58.3% (88/151) would administer sodium bicarbonate in a cardiac arrest scenario (p=0.004). 47.3% (71/150) selected a pH threshold at or below which they would administer sodium bicarbonate (mean, 6.94±0.013; median, 7.00; range, 6.50-7.20; interquartile range, 6.90-7.00), whereas 20.5% (31/151) selected a base excess threshold (mean, -15.62±0.78; median, -16; range, -20 to -4; interquartile range, -20 to -14). Both pH and duration of resuscitation were strongly associated with the decision to administer sodium bicarbonate (p<0.0001). Respondents' perceptions regarding a colleague's likelihood of administering sodium bicarbonate to the same patient under the same circumstances reflect an acknowledgment of disparate practices with respect to sodium bicarbonate use. 53.0% (79/149) felt current American Heart Association guidelines help them in deciding whether to administer sodium bicarbonate to critically ill patients, and 84% would support a randomized trial. Differences of opinion exist among pediatric acute care physicians with respect to the timing and appropriateness of sodium bicarbonate administration during resuscitation. Most indicated they would support moving forward with a clinical trial.
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%