Merck
  • Directional synthesis of ethylbenzene through catalytic transformation of lignin.
Directional synthesis of ethylbenzene through catalytic transformation of lignin.
Bioresource technology (2013-06-20)
Minghui Fan, Peiwen Jiang, Peiyan Bi, Shumei Deng, Lifeng Yan, Qi Zhai, Tiejun Wang, Quanxin Li
ABSTRACT
Transformation of lignin to ethylbenzene can provide an important bulk raw material for the petrochemical industry. This work explored the production of ethylbenzene from lignin through the directional catalytic depolymerization of lignin into the aromatic monomers followed by the selective alkylation of the aromatic monomers. For the first step, the aromatics selectivity of benzene derived from the catalytic depolymerization of lignin reached about 90.2 C-mol% over the composite catalyst of Re-Y/HZSM-5 (25). For the alkylation of the aromatic monomers in the second step, the highest selectivity of ethylbenzene was about 72.3 C-mol% over the HZSM-5 (25) catalyst. The reaction pathway for the transformation of lignin to ethylbenzene was also addressed. Present transformation potentially provides a useful approach for the production of the basic petrochemical material and development of high-end chemicals utilizing lignin as the abundant natural aromatic resource.
Product Description
Supelco
Ethylbenzene, analytical standard
Sigma-Aldrich
Ethylbenzene, anhydrous, 99.8%
Sigma-Aldrich
Ethylbenzene, ReagentPlus®, 99%
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 1 %, NMR tube size 5 mm × 7 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), chromium(III) acetylacetonate 0.1 mg/mL, TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.
Supelco
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 8 mm × 8 in.
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzene solution, suitable for NMR (reference standard), 1% in chloroform-d (99.8 atom % D)