Merck
Evolution of the salivary apyrases of blood-feeding arthropods.
Gene (2013-06-25)
Austin L Hughes
ABSTRACT
Phylogenetic analyses of three families of arthropod apyrases were used to reconstruct the evolutionary relationships of salivary-expressed apyrases, which have an anti-coagulant function in blood-feeding arthropods. Members of the 5'nucleotidase family were recruited for salivary expression in blood-feeding species at least five separate times in the history of arthropods, while members of the Cimex-type apyrase family have been recruited at least twice. In spite of these independent events of recruitment for salivary function, neither of these families showed evidence of convergent amino acid sequence evolution in salivary-expressed members. On the contrary, in the 5'-nucleotide family, salivary-expressed proteins conserved ancestral amino acid residues to a significantly greater extent than related proteins without salivary function, implying parallel evolution by conservation of ancestral characters. This unusual pattern of sequence evolution suggests the hypothesis that purifying selection favoring conservation of ancestral residues is particularly strong in salivary-expressed members of the 5'-nucleotidase family of arthropods because of constraints arising from expression within the vertebrate host.
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder