Merck
  • Spectroscopic and electrochemical characterization of some Schiff base metal complexes containing benzoin moiety.
Spectroscopic and electrochemical characterization of some Schiff base metal complexes containing benzoin moiety.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-06-25)
M S El-Shahawi, M S Al-Jahdali, A S Bashammakh, A A Al-Sibaai, H M Nassef
ABSTRACT
The ligation behavior of bis-benzoin ethylenediamine (B2ED) and benzoin thiosemicarbazone (BTS) Schiff bases towards Ru(3+), Rh(3+), Pd(2+), Ni(2+) and Cu(2+) were determined. The bond length of M-N and spectrochemical parameters (10Dq, β, B and LFSE) of the complexes were evaluated. The redox characteristics of selected complexes were explored by cyclic voltammetry (CV) at Pt working electrode in non aqueous solvents. Au mesh (100 w/in.) optically transparent thin layer electrode (OTTLE) was also used for recording thin layer CV for selected Ru complex. Oxidation of some complexes occurs in a consecutive chemical reaction of an EC type mechanism. The characteristics of electron transfer process of the couples M(2+)/M(3+) and M(3+)/M(4+) (M=Ru(3+), Rh(3+)) and the stability of the complexes towards oxidation and/or reduction were assigned. The nature of the electroactive species and reduction mechanism of selected electrode couples were assigned.
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Supelco
Ethylenediamine, analytical standard
Sigma-Aldrich
Ethylenediamine, puriss. p.a., absolute, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
Ethylenediamine, BioXtra
Sigma-Aldrich
Ethylenediamine, meets USP testing specifications
Sigma-Aldrich
Benzoin, 98%
Sigma-Aldrich
Benzoin, purified by sublimation, ≥99.5%
Sigma-Aldrich
Ethylenediamine dihydrochloride, 98%