Merck
  • Sarcosine and other metabolites along the choline oxidation pathway in relation to prostate cancer--a large nested case-control study within the JANUS cohort in Norway.
Sarcosine and other metabolites along the choline oxidation pathway in relation to prostate cancer--a large nested case-control study within the JANUS cohort in Norway.
International journal of cancer (2013-06-26)
Stefan de Vogel, Arve Ulvik, Klaus Meyer, Per Magne Ueland, Ottar Nygård, Stein Emil Vollset, Grethe S Tell, Jesse F Gregory, Steinar Tretli, Tone Bjørge
ABSTRACT
Methyl group donors and intermediates of one-carbon metabolism affect DNA synthesis and DNA methylation, and may thereby affect prostate carcinogenesis. Choline, the precursor of betaine, and the one-carbon metabolite sarcosine have been associated with increased prostate cancer risk. Within JANUS, a prospective cohort in Norway (n = 317,000) with baseline serum samples, we conducted a nested case-control study among 3,000 prostate cancer cases and 3,000 controls. Using conditional logistic regression, odds ratios (ORs) and 95% confidence intervals (CIs) for prostate cancer risk were estimated according to quintiles of circulating betaine, dimethylglycine (DMG), sarcosine, glycine and serine. High sarcosine and glycine concentrations were associated with reduced prostate cancer risk of borderline significance (sarcosine: highest vs. lowest quintile OR = 0.86, CI = 0.72-1.01, p(trend) = 0.03; glycine: OR = 0.83, CI = 0.70-1.00, p(trend) = 0.07). Serum betaine, DMG and serine were not associated with prostate cancer risk. However, individuals with a high glycine/serine ratio were at decreased prostate cancer risk (OR = 0.74, CI = 0.69-0.85, p(trend) < 0.001). This population-based study suggested that men with high serum sarcosine or glycine concentrations have modestly reduced prostate cancer risk. Ratios of metabolites reflecting one-carbon balance may be associated with prostate cancer risk, as demonstrated for the glycine/serine ratio, and should be explored in future studies.
Sigma-Aldrich
Choline chloride, ≥99%
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
N,N-Dimethylglycine, ≥99%
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Sarcosine, crystallized, ≥98.0% (T)
Sigma-Aldrich
Sarcosine, BioXtra
Sigma-Aldrich
Sarcosine, 98%