Merck
  • A cytometric bead assay for sensitive DNA detection based on enzyme-free signal amplification of hybridization chain reaction.
A cytometric bead assay for sensitive DNA detection based on enzyme-free signal amplification of hybridization chain reaction.
Biosensors & bioelectronics (2013-06-29)
Wei Ren, Hongmei Liu, Wenxia Yang, Yunlong Fan, Lang Yang, Yucong Wang, Chenghui Liu, Zhengping Li
ABSTRACT
A versatile flow cytometric bead assay (CBA) is developed for sensitive DNA detection by integrating the advantages of hybridization chain reaction (HCR) for enzyme-free signal amplification, flow cytometry for robust and rapid signal readout as well as magnetic beads (MBs) for facile separation. In this HCR-CBA, a biotinylated hairpin DNA (Bio-H1) is firstly immobilized on streptavidin-functionalized MBs. Upon the addition of target DNA, each target would hybridize with one Bio-H1 to open its hairpin structure and subsequently initiate a cascade of hybridization events between two species of fluorescent DNA hairpin probes (H1*/H2*) to form a nicked double helical DNA structure, resulting in amplified accumulation of numerous fluorophores on the MBs. Finally, the fluorescent MBs are directly analyzed by flow cytometry. This technique enables quantitative analysis of the HCR products anchored on the MBs as a function of target DNA concentration, and analysis of each sample can be completed within few minutes. Therefore, the HCR-CBA approach provides a practical DNA assay with greatly improved sensitivity. The detection limit of a model DNA target is 0.5 pM (3σ), which is about 3 orders of magnitude lower compared with traditional hybridization methods without HCR. Furthermore, the signal of complementary target can be clearly distinguished from that of single-base mismatched sequences, indicating the high specificity of the HCR-CBA. Moreover, this strategy is also successfully applied to the DNA analysis in complex biological samples, showing great potential in gene analysis and disease diagnosis in clinical samples.
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Biotin, European Pharmacopoeia (EP) Reference Standard