Merck
  • Characterization of a bZIP gene highly expressed during ripening of the peach fruit.
Characterization of a bZIP gene highly expressed during ripening of the peach fruit.
Plant physiology and biochemistry : PPB (2013-07-13)
Alessandro Lovisetto, Flavia Guzzo, Alice Tadiello, Enrico Confortin, Anna Pavanello, Alessandro Botton, Giorgio Casadoro
ABSTRACT
A ripening specific bZIP gene of peach was studied by ectopically expressing it in tomato. Two lines, with either a mild or a strong phenotype, respectively, were analyzed in detail. Transgenic fruit morphology was normal, yet the time spent to proceed through the various ripening stages was longer compared to wild type. In agreement with this finding the transgenic berries produced less ethylene, and also had a modified expression of some ripening-related genes that was particularly evident in berries with a strong phenotype. In particular, in the latter fruits polygalacturonase and lipoxygenase genes, but also genes coding for transcription factors (TFs) important for tomato ripening (i.e. TAGL1, CNR, APETALA2a, NOR) did not show the expected decreased expression in the red berries. As regards the RIN gene, its expression continued to increase in both mild and strong lines, and this is in agreement with the dilated ripening times. Interestingly, a metabolomic analysis of berries at various stages of ripening showed that the longer time spent by the transgenic berries to proceed from a stage to another was not due to a slackened metabolism. In fact, the differences in amount of stage-specific marker metabolites indicated that the transgenic berries had a very active metabolism. Therefore, the dilated ripening and the enhanced metabolism of the berries over-expressing the bZIP gene suggest that such gene might regulate ripening by acting as a pacemaker for some of the ripening metabolic pathways.
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type I-B, lyophilized powder, ≥50,000 units/mg solid
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type V, ammonium sulfate suspension, 500,000-1,000,000 units/mg protein
Sigma-Aldrich
Pectinase from Aspergillus niger, BioReagent, suitable for plant cell culture, aqueous glycerol solution, ≥5 units/mg protein (Lowry)
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, lyophilized powder, ≥0.3 units/mg solid
Sigma-Aldrich
Pectolyase from Aspergillus japonicus, suitable for plant cell culture, lyophilized powder
Sigma-Aldrich
Pectinase from Rhizopus sp., powder, 400-800 units/g solid
Sigma-Aldrich
Ethylene, ≥99.5%