Merck
  • Imatinib mesylate (Gleevec) induces human corpus cavernosum relaxation by inhibiting receptor tyrosine kinases (RTKs): identification of new RTK targets.
Imatinib mesylate (Gleevec) induces human corpus cavernosum relaxation by inhibiting receptor tyrosine kinases (RTKs): identification of new RTK targets.
Urology (2013-07-17)
Serap Gur, Suresh C Sikka, Asim B Abdel-Mageed, Zakaria Y Abd Elmageed, Bashir Rezk, Edward Pankey, Philip J Kadowitz, Wayne J G Hellstrom
ABSTRACT
To evaluate the effect of the tyrosine kinase inhibitor imatinib mesylate (Gleevec) on human corpus cavernosum (HCC) smooth muscle tone. HCC were obtained from 18 erectile dysfunction (ED) patients undergoing penile prosthesis surgery. The effects of imatinib in HCC strips were investigated in the presence of various inhibitors. The human phosphoreceptor protein tyrosine kinase (PTK) array (Proteome Profiler Array) detected changes in receptor phosphorylation before and after imatinib. Immunohistochemistry was used to localize phosphorylated c-kit (CD117/stem cell factor) in HCC smooth muscle cells. Phenylephrine-induced contraction in HCC was significantly inhibited by imatinib (97.7% ± 2.3%). l-nitro-arginine methyl ester (l-NAME) or guanylyl cyclase inhibitor [1H-1,2,4] oxadiazolo [4,3-a]quinoxalin-1-one (ODQ) alone did not reverse the effect of imatinib, but suppressed this response in combination (18.0% ± 0.6%). The K(+) channel blockers (apamin and tetraethyl ammonium) decreased the imatinib-induced relaxation by 64% and 51%, respectively. PTK microarray analysis of 42 different phospho-receptor tyrosine kinases showed 14 were clearly activated in HCC. Imatinib treatment significantly inhibited phosphorylation of PTKs. A high level of CD117/c-kit-positive immunostaining was detected in untreated HCC smooth muscle, but not in treated HCC. Imatinib caused HCC smooth muscle relaxation in vitro mediated by nitric oxide/guanosine monophosphate signaling, involving the large-conductance Ca(2+)-activated K(+)-channels (BK(Ca)) or by inhibiting the upregulated PTK pathway. These results suggest that imatinib may also benefit erectile dysfunction patients who are not responsive to phosphodiesterase-5 inhibitors.
Sigma-Aldrich
Tetraethylammonium chloride hydrate
Sigma-Aldrich
Tetraethylammonium iodide, 98%
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 35 wt. % in H2O
Sigma-Aldrich
Apamin, synthetic, ≥97% (HPLC)
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)
Sigma-Aldrich
Tetraethylammonium chloride, ≥98% (titration)
Sigma-Aldrich
Tetraethylammonium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Supelco
Tetraethylammonium chloride, for electrochemical analysis, ≥99.0%
Supelco
Tetraethylammonium hydroxide solution, ~1.0 M (CH3CH2)4NOH in H2O, electrochemical grade
Sigma-Aldrich
Tetraethylammonium hydroxide solution, ~25% in methanol (~1.5 M)
Sigma-Aldrich
Tetraethylammonium hydroxide solution, 20 wt. % in H2O