Merck
  • The identification of degradation products and degradation pathway of malvidin-3-glucoside and malvidin-3,5-diglucoside under microwave treatment.
The identification of degradation products and degradation pathway of malvidin-3-glucoside and malvidin-3,5-diglucoside under microwave treatment.
Food chemistry (2013-07-23)
Mengyao Zhao, Yinghua Luo, Yuan Li, Xin Liu, Jihong Wu, Xiaojun Liao, Fang Chen
ABSTRACT
Understanding the degradation of anthocyanins (Acys) during microwave treatment is required for the application of this novel technology to fruit and vegetable processing. The pH 3.5 buffer solutions containing 50mg/l malvidin-3-glucoside (Mv-3-glu) and malvidin-3,5-diglucoside (Mv-3,5-diglu), respectively, were treated with microwaves at 700 W. In comparison, the samples were heated at 98±2 °C in a water bath. In total, nine degradation products were detected (using LC-UV/Vis-MS/MS) for each Acys after microwave treatment. Anthocyanone A, hydroxycoumarins and dihydroxy phenylacetaldehyde were characterised as the main degradation products of both Acys under microwave conditions, which were not detected in the thermal treatment. In addition, syringic acid, 2,4,6-trihydroxybenzaldehyde, coumarin glucosides and 2,6-dimethoxyphenol were found simultaneously under the microwave and thermal treatments. During the microwave treatment, hydrogen peroxide was formed and resulted in the Baeyer-Villiger-type oxidation of the Acys, which was the main degradation pathway. At the same time, the conventional thermal degradation pathway of Acys also occurred.
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in isopropanol
Sigma-Aldrich
Acetaldehyde solution, 40 wt. % in H2O
Sigma-Aldrich
Phenylacetaldehyde, ≥90%
Sigma-Aldrich
Acetaldehyde solution, natural, 50 wt. % ethanol, FG
Sigma-Aldrich
Acetaldehyde solution, 50 wt. % in ethanol
Sigma-Aldrich
Phenylacetaldehyde, ≥95%, FCC, FG
Sigma-Aldrich
Acetaldehyde solution, 5 M in THF
Sigma-Aldrich
Acetaldehyde, ACS reagent, ≥99.5%
Sigma-Aldrich
Oenin chloride, ≥90% (HPLC)
Sigma-Aldrich
Acetaldehyde, natural, FG
Supelco
Acetaldehyde, PESTANAL®, analytical standard
Sigma-Aldrich
Acetaldehyde, puriss. p.a., anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Acetaldehyde, ReagentPlus®, ≥99.0% (GC)