- Phenylalanine-containing cyclic dipeptides--the lowest molecular weight hydrogelators based on unmodified proteinogenic amino acids.
Phenylalanine-containing cyclic dipeptides--the lowest molecular weight hydrogelators based on unmodified proteinogenic amino acids.
Chemical communications (Cambridge, England) (2013-07-26)
Alexander J Kleinsmann, Boris J Nachtsheim
PMID23884374
ABSTRACT
Cyclic dipeptides (diketopiperazines - DKPs) that are based on the proteinogenic amino acid phenylalanine in combination with serine, cysteine, glutamate, histidine and lysine are described as simple and remarkable low molecular weight hydrogelators. Blends of selected DKPs show remarkable pH-dependent properties and can be applied as easy to tune materials in drug delivery.
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor