Merck
  • Neurotoxic effect of subacute benzo(a)pyrene exposure on gene and protein expression in Sprague-Dawley rats.
Neurotoxic effect of subacute benzo(a)pyrene exposure on gene and protein expression in Sprague-Dawley rats.
Environmental toxicology and pharmacology (2013-07-31)
Shu-qun Cheng, Yin-yin Xia, Jun-lin He, Xue-qing Liu, Xue-mei Chen, Yu-bin Ding, Ying-xiong Wang, Bing Peng, Bai-jie Tu
ABSTRACT
Benzo(a)pyrene (B[a]P) is an environmental carcinogen that induces tumors in many animal species, but the neurotoxic effects of B[a]P have not been well studied. In the present study, we investigated the effects of subacute exposure to B[a]P in Sprague-Dawley (SD) rats. Male rats received daily injections of either B[a]P (0, 1, 2.5, or 6.25mg/kg, i.p.) or vehicle for 45 days. Exposure to B[a]P affected the behavior of rats in the Morris water maze test. Gene microarray and real-time PCR analyses revealed that exposure to B[a]P affected signal transduction in the rat hippocampus. Protein microarray analysis revealed that altered protein expression played a role in cell death in the functional annotation cluster analysis. Finally, major vault protein was found to display low cDNA and protein expression levels. The present study explored some of the possible mechanisms underlying B[a]P neurotoxicity and provided evidence that B[a]P plays a neurotoxic role in rats.
Product Description
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)