Merck
Protein:protein interactions in control of a transcriptional switch.
Journal of molecular biology (2013-07-31)
Poorni R Adikaram, Dorothy Beckett
ABSTRACT
Protein partner exchange plays a key role in regulating many biological switches. Although widespread, the mechanisms dictating protein partner identity and, therefore, the outcome of a switch have been determined for a limited number of systems. The Escherichia coli protein BirA undergoes a switch between posttranslational biotin attachment and transcription repression in response to cellular biotin demand. Moreover, the functional switch reflects formation of alternative mutually exclusive protein:protein interactions by BirA. Previous studies provided a set of alanine-substituted BirA variants with altered kinetic and equilibrium parameters of forming these interactions. In this work, DNase I footprinting measurements were employed to investigate the consequences of these altered properties for the outcome of the BirA functional switch. The results support a mechanism in which BirA availability for DNA binding and, therefore, transcription repression is controlled by the rate of the competing protein:protein interaction. However, occupancy of the transcriptional regulatory site on DNA by BirA is exquisitely tuned by the equilibrium constant governing its homodimerization.
Product Description
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Hydroquinidine, 95%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Sigma-Aldrich
Tetramethylammonium iodide, 99%
Sigma-Aldrich
Tetramethylammonium chloride solution, Molecular Biology
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Biotin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%