Merck
  • Phosphodiesterase 5 inhibitor acts as a potent agent sensitizing acute myeloid leukemia cells to 67-kDa laminin receptor-dependent apoptosis.
Phosphodiesterase 5 inhibitor acts as a potent agent sensitizing acute myeloid leukemia cells to 67-kDa laminin receptor-dependent apoptosis.
FEBS letters (2013-08-07)
Motofumi Kumazoe, Yoonhee Kim, Jaehoon Bae, Mika Takai, Motoki Murata, Yumi Suemasu, Kaori Sugihara, Shuya Yamashita, Shuntaro Tsukamoto, Yuhui Huang, Kanami Nakahara, Koji Yamada, Hirofumi Tachibana
ABSTRACT
(-)-Epigallocatechin-3-O-gallate (EGCG), a polyphenol in green tea, induces apoptosis in acute myeloid leukemia (AML) cells without affecting normal cells. In this study, we observed that cGMP acts as a cell death mediator of the EGCG-induced anti-AML effect through acid sphingomyelinase activation. EGCG activated the Akt/eNOS axis, a well-known mechanism in vascular cGMP upregulation. We also observed that a major cGMP negative regulator, phosphodiesterase 5, was overexpressed in AML cells, and PDE5 inhibitor, an anti-erectile dysfunction drug, synergistically enhanced the anti-AML effect of EGCG. This combination regimen killed AML cells via overexpressed 67-kDa laminin receptors.
Sigma-Aldrich
Sphingomyelinase from Staphylococcus aureus, buffered aqueous glycerol solution, 100-300 units/mg protein (Lowry)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
Sphingomyelinase from Bacillus cereus, buffered aqueous glycerol solution, ≥100 units/mg protein (Lowry)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Sphingomyelinase from Bacillus cereus, lyophilized powder, ≥100 units/mg protein
Supelco
(−)-Epigallocatechin gallate, analytical standard
Epigallocatechin gallate, primary reference standard