Merck
  • Total synthesis of enantiopure pyrrhoxanthin: alternative methods for the stereoselective preparation of 4-alkylidenebutenolides.
Total synthesis of enantiopure pyrrhoxanthin: alternative methods for the stereoselective preparation of 4-alkylidenebutenolides.
Chemistry (Weinheim an der Bergstrasse, Germany) (2013-08-14)
Belén Vaz, Leticia Otero, Rosana Álvarez, Ángel R de Lera
ABSTRACT
A new stereocontrolled total synthesis of the configurationally labile C37 -norcarotenoid pyrrhoxanthin in enantiopure form has been completed. A highly stereoselective Horner-Wadsworth-Emmons (HWE) condensation of a C17-allylphosphonate and a C20-aldehyde was used as the last conjunctive step. Both a Sonogashira reaction to form the C17-phosphonate and the final HWE condensation proved to be compatible with the sensitive C7-C10 enyne E configuration. Regioselective (5-exo-dig) silver-promoted lactonization reactions of three alternative pent-2-en-4-ynoic acid precursors with increased complexity, including a fully functionalized C20-fragment, were explored for the preparation of the γ-alkylidenebutenolide fragment. This survey extends the existing methodologies for the preparation of oxygen-containing carotenoids (xanthophylls) and streamlines the synthesis of additional members of the C37-norcarotenoid butenolide family of natural products.
Sigma-Aldrich
2(5H)-Furanone, 98%
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%