Merck
  • Phosphorylation of Noxo1 at threonine 341 regulates its interaction with Noxa1 and the superoxide-producing activity of Nox1.
Phosphorylation of Noxo1 at threonine 341 regulates its interaction with Noxa1 and the superoxide-producing activity of Nox1.
The FEBS journal (2013-08-21)
Asataro Yamamoto, Ryu Takeya, Masaki Matsumoto, Keiichi I Nakayama, Hideki Sumimoto
ABSTRACT
Superoxide production by Nox1, a member of the Nox family NAPDH oxidases, requires expression of its regulatory soluble proteins Noxo1 (Nox organizer 1) and Noxa1 (Nox activator 1) and is markedly enhanced upon cell stimulation with phorbol 12-myristate 13-acetate (PMA), a potent activator of protein kinase C (PKC). The mechanism underlying PMA-induced enhancement of Nox1 activity, however, remains to be elucidated. Here we show that, in response to PMA, Noxo1 undergoes phosphorylation at multiple sites, which is inhibited by the PKC inhibitor GF109203X. Among them, Thr341 in Noxo1 is directly phosphorylated by PKC in vitro, and alanine substitution for this residue reduces not only PMA-induced Noxo1 phosphorylation but also PMA-dependent enhancement of Nox1-catalyzed superoxide production. Phosphorylation of Thr341 allows Noxo1 to sufficiently interact with Noxa1, an interaction that participates in Nox1 activation. Thus phosphorylation of Noxo1 at Thr341 appears to play a crucial role in PMA-elicited activation of Nox1, providing a molecular link between PKC-mediated signal transduction and Nox1-catalyzed superoxide production. Furthermore, Ser154 in Noxo1 is phosphorylated in both resting and PMA-stimulated cells, and the phosphorylation probably participates in a PMA-independent constitutive activity of Nox1. Ser154 may also be involved in protein kinase A (PKA) mediated regulation of Nox1; this serine is the major residue that is phosphorylated by PKA in vitro. Thus phosphorylation of Noxo1 at Thr341 and at Ser154 appears to regulate Nox1 activity in different manners. Noxo1 binds to p22phox by pull down (1, 2, 3) Noxo1 binds to Noxo1 by pull down (View interaction) Noxa1 binds to Noxo1 by pull down (1, 2, 3, 4, 5).
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
4α-Phorbol 12-myristate 13-acetate, solid, ≥95% (TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology