Merck
  • Incremental cost-effectiveness of pharmacotherapy and two brief cognitive-behavioral therapies compared with usual care for panic disorder and noncardiac chest pain.
Incremental cost-effectiveness of pharmacotherapy and two brief cognitive-behavioral therapies compared with usual care for panic disorder and noncardiac chest pain.
The Journal of nervous and mental disease (2013-09-03)
Joannie Poirier-Bisson, André Marchand, Marie-Eve Pelland, Marie-Josée Lessard, Gilles Dupuis, Richard Fleet, Pasquale Roberge
ABSTRACT
The aim of this study was to assess the incremental cost-effectiveness ratios (ICERs) of two brief cognitive-behavioral therapy (CBT)-based interventions and a pharmacological treatment, compared with usual care, initiated in the emergency department (ED) for individuals with panic disorder (PD) with a chief complaint of noncardiac chest pain. A total of 69 patients were followed up to 6 months. The primary outcome variables were direct and indirect costs of treatment and PD severity. Panic management (PM) had an ICER of $124.05, per the Anxiety Disorders Interview Schedule for Diagnostic and Statistical Manual of Mental Disorders, Fourth Edition, severity score change (95% confidence interval [CI], $54.63-$314.57), compared with pharmacotherapy (paroxetine), with an ICER of $213.90 (95% CI, $133.51-$394.94), and brief CBT, with an ICER of $309.31 (95% CI, $151.27-$548.28). The pharmacological and CBT interventions were associated with a greater clinical improvement compared with usual care at posttest. PM presented a superior ICER, suggesting that it may be a promising treatment option to implement in EDs.
Product Description
Paroxetine hydrochloride (anhydrous), European Pharmacopoeia (EP) Reference Standard
Supelco
Paroxetine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Paroxetine maleate salt, ≥98% (HPLC), solid