Merck
  • Membrane protein stability depends on the concentration of compatible solutes--a single molecule force spectroscopic study.
Membrane protein stability depends on the concentration of compatible solutes--a single molecule force spectroscopic study.
Biological chemistry (2013-09-12)
Arpita Roychoudhury, Adeline Bieker, Dieter Häussinger, Filipp Oesterhelt
ABSTRACT
Compatible solutes are small, uncharged, zwitter ionic, osmotically active molecules produced and accumulated by microorganisms inside their cell to counteract different kinds of environmental stress. They enhance protein stability without interfering with the metabolic pathways even at molar concentrations. In this paper, we report the stabilizing effects of compatible solutes, ectoine, betaine and taurine on membrane protein bacteriorhodopsin at different concentrations. Using atomic force microscopy based single molecule force spectroscopy the impact of the osmolytes was quantified by measuring the forces required to pull the protein out of the membrane and the change in the persistence lengths of the unfolded polypeptide chain. Increase in unfolding forces were observed, indicating the strengthening of intramolecular interactions, which are vital for protein stability. The decrease in persistence lengths was recorded and showed increasing tendencies of the polypeptide strand to coil up. Interestingly, it was revealed that these molecules have different stabilizing effects on protein unfolding at different concentrations. The results show that the unfolding of single protein provides insight to the structure-dynamic relationship between the protein and compatible solute molecules at sub-nanometer scale. This also helps to understand the molecular mechanism involved in protein stabilization by organic osmolytes.
Sigma-Aldrich
Betaine, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Taurine, ≥98%, FG
Supelco
Taurine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Betaine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Taurine, ≥99%
Sigma-Aldrich
Taurine, suitable for cell culture, meets USP testing specifications
Sigma-Aldrich
Betaine hydrochloride, ≥99%
Sigma-Aldrich
Ectoine, ≥95.0% (HPLC)
SAFC
Taurine
Sigma-Aldrich
Taurine, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ectoine, osmoprotectant
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent