Merck
  • Mutation of Trp(93) of MauG to tyrosine causes loss of bound Ca(2+) and alters the kinetic mechanism of tryptophan tryptophylquinone cofactor biosynthesis.
Mutation of Trp(93) of MauG to tyrosine causes loss of bound Ca(2+) and alters the kinetic mechanism of tryptophan tryptophylquinone cofactor biosynthesis.
The Biochemical journal (2013-09-13)
Sooim Shin, Manliang Feng, Victor L Davidson
ABSTRACT
The dihaem enzyme MauG catalyses a six-electron oxidation required for post-translational modification of preMADH (precursor of methylamine dehydrogenase) to complete the biosynthesis of its TTQ (tryptophan tryptophylquinone) cofactor. Trp93 of MauG is positioned midway between its two haems, and in close proximity to a Ca2+ that is critical for MauG function. Mutation of Trp93 to tyrosine caused loss of bound Ca2+ and changes in spectral features similar to those observed after removal of Ca2+ from WT (wild-type) MauG. However, whereas Ca2+-depleted WT MauG is inactive, W93Y MauG exhibited TTQ biosynthesis activity. The rate of TTQ biosynthesis from preMADH was much lower than that of WT MauG and exhibited highly unusual kinetic behaviour. The steady-state reaction exhibited a long lag phase, the duration of which was dependent on the concentration of preMADH. The accumulation of reaction intermediates, including a diradical species of preMADH and quinol MADH (methylamine dehydrogenase), was detected during this pre-steady-state phase. In contrast, steady-state oxidation of quinol MADH to TTQ, the final step of TTQ biosynthesis, exhibited no lag phase. A kinetic model is presented to explain the long pre-steady-state phase of the reaction of W93Y MauG, and the role of this conserved tryptophan residue in MauG and related dihaem enzymes is discussed.
Product Description
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)