Merck
Lhx3-Chx10 reticulospinal neurons in locomotor circuits.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-09-13)
Frédéric Bretzner, Robert M Brownstone
ABSTRACT
Motor behaviors result from the interplay between the brain and the spinal cord. Reticulospinal neurons, situated between the supraspinal structures that initiate motor movements and the spinal cord that executes them, play key integrative roles in these behaviors. However, the molecular identities of mammalian reticular formation neurons that mediate motor behaviors have not yet been determined, thus limiting their study in health and disease. In the medullary reticular formation of the mouse, we identified neurons that express the transcription factors Lhx3 and/or Chx10, and demonstrate that these neurons form a significant component of glutamatergic reticulospinal pathways. Lhx3-positive medullary reticular formation neurons express Fos following a locomotor task in the adult, indicating that they are active during walking. Furthermore, they receive functional inputs from the mesencephalic locomotor region and have electrophysiological properties to support tonic repetitive firing, both of which are necessary for neurons that mediate the descending command for locomotion. Together, these results suggest that Lhx3/Chx10 medullary reticular formation neurons are involved in locomotion.
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Sigma-Aldrich
Biotin, ≥99.0% (T)
Sigma-Aldrich
Biotin, meets USP testing specifications
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Biotin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran, enzymatic synth.
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran solution from Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, Mw 1,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 12,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 670,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 410,000
Supelco
Dextran, analytical standard, 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 50,000
Supelco
Dextran, analytical standard, 670,000
Supelco
Dextran, analytical standard, 270,000
Supelco
Dextran, analytical standard, 410,000
Supelco
Dextran, analytical standard, 150,000