Merck
  • The presence of heat-labile factors interfering with binding analysis of fibrinogen with ferritin in horse plasma.
The presence of heat-labile factors interfering with binding analysis of fibrinogen with ferritin in horse plasma.
Acta veterinaria Scandinavica (2013-09-24)
Kazuma Takahashi, Takashi Kondo, Yasunaga Yoshikawa, Kiyotaka Watanabe, Koichi Orino
ABSTRACT
Horse fibrinogen has been identified as a plasma specific ferritin-binding protein. There are two ways in the binding of ferritin-binding protein with ferritin: one is direct binding and the other is indirect binding which is heme-mediated. The aim of this study was to analyze the binding between horse fibrinogen and ferritin. Although fibrinogen in horse plasma did not show the binding to ferritin coated on the plate wells, after following heat-treatment (60°C, 30 min) of horse plasma, plasma fibrinogen as well as purified horse fibrinogen bound to plates coated with horse spleen ferritin, but not with its apoferritin which lost heme as well as iron after the treatment of reducing reagent. Binding of purified or plasma fibrinogen to ferritin was inhibited by hemin and Sn-protoporphyrin IX (Sn-PPIX), but not by PPIX or Zn-PPIX. Heat-treatment of horse plasma enabled plasma fibrinogen to bind to plate well coated with holo-ferritin. From the binding analysis of fibrinogen and ferritin, it is suggested that horse fibrinogen recognized iron or tin in complexed with the heme- or the hemin-ring, and also suggest that some fibrinogens circulate in the form of a complex with ferritin and/or heat-labile factors which inhibit the binding of fibrinogen with ferritin.
Sigma-Aldrich
Tin, powder, -100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Tin, powder, 10 μm, 99% trace metals basis
Sigma-Aldrich
Tin, ≥99%, powder
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Tin, foil, thickness 0.127 mm, 99.9%
Sigma-Aldrich
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sigma-Aldrich
Tin, powder, <150 μm, 99.5% trace metals basis
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sigma-Aldrich
Tin, 99.8%, shot, 3 mm
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
Tin, shot, 99.999% trace metals basis
Sigma-Aldrich
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis