Merck
  • Efficacy of urine alkalinization by oral administration of sodium bicarbonate: a prospective open-label trial.
Efficacy of urine alkalinization by oral administration of sodium bicarbonate: a prospective open-label trial.
The American journal of emergency medicine (2013-09-24)
Barak Cohen, Ido Laish, Tal Brosh-Nissimov, Azik Hoffman, Lior H Katz, Rony Braunstein, Ram Sagi, Goldman Michael
ABSTRACT
Urine alkalinization is indicated for various medical conditions. Alkalinization is usually achieved by intravenous administration of alkali substances titrated by repeated urinalyses. Some situations such as mass casualty events might require urine alkalinization by the oral route. We evaluated the efficacy of oral sodium bicarbonate administration for urine alkalinization. In a prospective open-label trial, 4 g of sodium bicarbonate was administered orally 3 times daily to 9 healthy volunteers for 24 hours. Serial blood and urine samples were collected, and urine pH was evaluated. Plasma electrolytes and pH were also measured for safety purposes. All participants had a urine pH of at least 7 after 10 hours. At 20 hours, all participants had a urine pH of at least 8. No adverse effects or abnormal blood results were documented during the 24-hour follow-up. Oral administration of a standard dose of sodium bicarbonate tablets resulted in effective urine alkalinization. Further research is needed to investigate the natural course of urine pH after cessation of our protocol and the efficacy of longer periods of treatment.
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C