Merck
A case of mannitol hypersensitivity.
Journal of pediatric hematology/oncology (2013-09-26)
Donita D Lightner, Kevin De Braganca, Stephen W Gilheeney, Yasmin Khakoo, Kim Kramer, Morad Balas
ABSTRACT
Mannitol is used for increased intracranial pressure and prevention of nephrotoxicity. We present a case report of a patient who experienced an anaphylactic response to mannitol and review the literature.
Product Description
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ACS reagent
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Diphenhydramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Diphenhydramine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Diphenhydramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)